Learn More
Benzyl 2,3-O-Isopropylidene-Alpha-D-mannofuranoside, TRC
Product Code. 30346571
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30346571 Supplier Toronto Research Chemicals Supplier No. TRCB2812001G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzyl 2,3-O-Isopropylidene-a-D-mannofuranoside
CAS 20689-03-6
Molecular Formula C16 H22 O6
Quantity 1 g
InChI Formula InChI=1S/C16H22O6/c1-16(2)21-13-12(11(18)8-17)20-15(14(13)22-16)19-9-10-6-4-3-5-7-10/h3-7,11-15,17-18H,8-9H2,1-2H3/t11-,12-,13+,14+,15+/m1/s1
SMILES CC1(C)O[C@@H]2[C@@H](OCc3ccccc3)O[C@H]([C@H](O)CO)[C@@H]2O1
IUPAC Name (1R)-1-[(3aS,4S,6R,6aS)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol
Molecular Weight (g/mol) 310.34
Formula Weight 310.1416