Learn More
Benzyl 2,3-O-Isopropylidene-Alpha-D-mannofuranoside, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl 2,3-O-Isopropylidene-a-D-mannofuranoside |
| CAS | 20689-03-6 |
| Molecular Formula | C16 H22 O6 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C16H22O6/c1-16(2)21-13-12(11(18)8-17)20-15(14(13)22-16)19-9-10-6-4-3-5-7-10/h3-7,11-15,17-18H,8-9H2,1-2H3/t11-,12-,13+,14+,15+/m1/s1 |
| SMILES | CC1(C)O[C@@H]2[C@@H](OCc3ccccc3)O[C@H]([C@H](O)CO)[C@@H]2O1 |
| IUPAC Name | (1R)-1-[(3aS,4S,6R,6aS)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol |
| Molecular Weight (g/mol) | 310.34 |
| Formula Weight | 310.1416 |