Learn More
Benzyl 2,3:5,6-Di-O-isopropylidene-Alpha-D-mannofuranoside, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl 2,3:5,6-Di-O-isopropylidene-a-D-mannofuranoside |
| CAS | 20689-02-5 |
| Molecular Formula | C19 H26 O6 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C19H26O6/c1-18(2)21-11-13(23-18)14-15-16(25-19(3,4)24-15)17(22-14)20-10-12-8-6-5-7-9-12/h5-9,13-17H,10-11H2,1-4H3/t13-,14-,15+,16+,17+/m1/s1 |
| SMILES | CC1(C)OC[C@@H](O1)[C@H]2O[C@H](OCc3ccccc3)[C@H]4OC(C)(C)O[C@@H]24 |
| IUPAC Name | (3aS,4S,6R,6aS)-6-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole |
| Molecular Weight (g/mol) | 350.41 |
| Formula Weight | 350.1729 |