Learn More
Benzoxazole, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 273-53-0 |
| Molecular Formula | C7H5NO |
| MDL Number | MFCD00005765 |
| Quantity | 5 g |
| SMILES | C1=CC=CC2=C1N=CO2 |
| Molecular Weight (g/mol) | 119.12 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzoxazole |