Learn More
Benzothiazole-6-carbonitrile, 98%
Product Code. 15405177
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15405177
Brand: Thermo Scientific Alfa Aesar H32454.06
Request Bulk or Custom Format
CAS: 58249-61-9 | C8H4N2S | 160.194 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58249-61-9
Molecular Formula C8H4N2S
Molecular Weight (g/mol) 160.194
MDL Number MFCD06659656
InChI Key WYRRTJKOMZONQO-UHFFFAOYSA-N
Synonym 6-cyanobenzothiazole, benzothiazole-6-carbonitrile, benzo d thiazole-6-carbonitrile, 6-benzothiazolecarbonitrile, 6-benzo d thiazolecarbonitrile, 6-cyanobenzothiazol, 6-cyanobenzthiazole, 6-benzothiazolecarbonitrile 6ci,9ci
PubChem CID 10607144
IUPAC Name 1,3-benzothiazole-6-carbonitrile
SMILES C1=CC2=C(C=C1C#N)SC=N2
Specifications
Melting Point 137°C to 138°C
Quantity 5 g
UN Number UN3439
Formula Weight 160.2
Percent Purity 98%
Chemical Name or Material Benzothiazole-6-carbonitrile