missing translation for 'onlineSavingsMsg'
Learn More

Benzopurpurine 4B (Technical Grade), TRC

Product Code. 30339023
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30339023

Brand: Toronto Research Chemicals TRCB2058105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzopurpurine 4B (Technical Grade)
CAS 992-59-6
Molecular Formula C34H26N6Na2O6S2
Quantity 5 g
Synonym C.I. Direct Red 2, C.I. Direct Red 2, Disodium Salt, Amanil Purpurine 4B, Atul Direct Red 4B, Azamin 4B, Azocard Red 4B, Bencidal Purple 4B, Benzanil Purpurine 4B, Benzopurpurin 4B, Benzopurpurin B, Benzopurpurine 4BKX, Benzopurpurine 4BX, Brasilamina Red 4B, C.I. 23500, Calcomine Red 4BX, Chrome Leather Red 4B, Cotton Red 4B, Diacotton Benzopurpurine 4B, Diamine Purpurine 4B, Diaphtamine, Diazamine Purpurine 4B, Diazine Red 4B, Diazol Purpurine 4B, Diphenyl Red 4B, Diphenyl Red 4BS, Direct Purpurine 4B, Direct Purpurine M 4B, Direct Red 2, Direct Red 4A, Direct Red 4B, Direct Red DCB, Direct Scarlet 4BE, Eclipse Red, Erie Benzo 4BP, Erie Red 4B, Fast Scarlet, Hispamin Red 4B, Kayaku Benzopurpurine 4B, Mitsui Benzopurpurine 4BX, Paper Red 4BS, Phenamine Purpurine 4B, Purpurin 4B, Purpurine 4B, Red 4B, Tertrodirect Red 4B
InChI Formula InChI=1S/C34H28N6O6S2.2Na/c1-19-15-21(11-13-27(19)37-39-29-17-31(47(41,42)43)23-7-3-5-9-25(23)33(29)35)22-12-14-28(20(2)16-22)38-40-30-18-32(48(44,45)46)24-8-4-6-10-26(24)34(30)36;;/h3-18H,35-36H2,1-2H3,(H,41,42,43)(H,44,45,46);;/q;2*+1/p-2
SMILES [Na+].[Na+].Cc1cc(ccc1N=Nc2cc(c3ccccc3c2N)S(=O)(=O)[O-])c4ccc(N=Nc5cc(c6ccccc6c5N)S(=O)(=O)[O-])c(C)c4
IUPAC Name disodium;4-amino-3-[(E)-[4-[4-[(E)-(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]-2-methylphenyl]diazenyl]naphthalene-1-sulfonate
Molecular Weight (g/mol) 724.72
Formula Weight 724.1151
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.