missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic-3,5-d2 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Benzoic acid 3,5-d2 |
| CAS | 37960-84-2 |
| Molecular Formula | C7 2H2 H4 O2 |
| Quantity | 0.5 g |
| Synonym | 3,5-Dideuteriobenzoic acid, Benzoic-3,5-d2 acid, Benzoic Acid D2 (Phenyl D2), Benzoic acid D2 (phenyl D2) |
| InChI Formula | InChI=1 S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5 H,(H,8,9)/i2D,3 D |
| SMILES | [2 H]c1cc([2 H])cc(c1)C(=O)O |
| IUPAC Name | 3,5-dideuteriobenzoic acid |
| Molecular Weight (g/mol) | 124.13 |
| Formula Weight | 124.0493 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?