Learn More
Benzoic-3,5-d2 Acid, CDN
Product Code. 30309381
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309381 0.25 g 0.25g
30310481 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309381 Supplier CDN Supplier No. CDND64720.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzoic acid 3,5-d2
CAS 37960-84-2
Molecular Formula C7 2H2 H4 O2
Quantity 0.25 g
Synonym 3,5-Dideuteriobenzoic acid, Benzoic-3,5-d2 acid, Benzoic Acid D2 (Phenyl D2), Benzoic acid D2 (phenyl D2)
InChI Formula InChI=1 S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5 H,(H,8,9)/i2D,3 D
SMILES [2 H]c1cc([2 H])cc(c1)C(=O)O
IUPAC Name 3,5-dideuteriobenzoic acid
Molecular Weight (g/mol) 124.13
Formula Weight 124.0493 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity