Learn More
Benzocaine Methanethiosulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzocaine Methanethiosulfonate |
| CAS | 212207-24-4 |
| Molecular Formula | C10 H13 N O4 S2 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C10H13NO4S2/c1-17(13,14)16-7-6-15-10(12)8-2-4-9(11)5-3-8/h2-5H,6-7,11H2,1H3 |
| SMILES | CS(=O)(=O)SCCOC(=O)c1ccc(N)cc1 |
| IUPAC Name | 2-methylsulfonylsulfanylethyl 4-aminobenzoate |
| Molecular Weight (g/mol) | 275.34 |
| Formula Weight | 275.0286 |