Learn More
Benzo[b]thiophene-D6, TRC
Specifications
Specifications
| Chemical Name or Material | Benzo[b]thiophene-D6 |
| CAS | 1444206-31-8 |
| Molecular Formula | C8 D6 S |
| Quantity | 2.5 mg |
| Synonym | Benzo[b]thiophene-2,3,4,5,6,7-D6 |
| InChI Formula | InChI=1S/C8H6S/c1-2-4-8-7(3-1)5-6-9-8/h1-6H/i1D,2D,3D,4D,5D,6D |
| SMILES | [2H]c1sc2c([2H])c([2H])c([2H])c([2H])c2c1[2H] |
| IUPAC Name | 2,3,4,5,6,7-hexadeuterio-1-benzothiophene |
| Molecular Weight (g/mol) | 140.235 |
| Formula Weight | 140.057 |