Learn More
Benzo[b]furan-3-ylacetic acid, 97%, Thermo Scientific™
Product Code. 10422643
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10422643 1 g 1g
10650895 250 mg 250mg
10366284 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10422643 Supplier Thermo Scientific Maybridge Supplier No. CC40801DA
 Restricted Item
Estimated Shipment: 28-09-2026
Chemical Identifiers
CAS 64175-51-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
InChI Key QWMVFCMIUUHJDH-UHFFFAOYSA-N
Synonym 2-benzofuran-3-yl acetic acid, 3-benzofuranacetic acid, benzo b furan-3-ylacetic acid, 2-1-benzofuran-3-yl acetic acid, benzofuran-3-yl-acetic acid, benzofuran-3-acetic acid, 1-benzofuran-3-ylacetic acid, 2-3-benzofuranyl acetic acid, pubchem7012, benzofuran-3-ylacetic acid
PubChem CID 2762940
IUPAC Name 2-(1-benzofuran-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=CO2)CC(=O)O
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-3-ylacetic acid