Learn More
Benzo[b]fluoren-11-one, TRC
Specifications
Specifications
| Chemical Name or Material | Benzo[b]fluoren-11-one |
| CAS | 3074-03-1 |
| Molecular Formula | C17H10O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C17H10O/c18-17-14-8-4-3-7-13(14)15-9-11-5-1-2-6-12(11)10-16(15)17/h1-10H |
| SMILES | C1=CC2=CC3=C(C=C2C=C1)C(=O)C4=CC=CC=C43 |
| IUPAC Name | benzo[b]fluoren-11-one |
| Molecular Weight (g/mol) | 230.26 |
| Formula Weight | 230.0732 |