Learn More
Benzeneselenol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 645-96-5 |
| Molecular Formula | C6H6Se |
| MDL Number | MFCD00000001 |
| Quantity | 25 g |
| SMILES | [SeH]C1=CC=CC=C1 |
| Molecular Weight (g/mol) | 157.07 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzeneselenol |