Learn More
Wilmad-LabGlass™ Benchtop Spectrometer NMR Tubes
Product Code. 15806905 Shop All SP Scienceware Products
Click to view available options
Length (English):
7 in.
8 in.
Model:
WG-BTNMR-7
WG-BTNMR-7-25
WG-BTNMR-8
WG-BTNMR-8-25
Quantity:
25
150
Unit Size:
Pack of 150
Pack of 25
Product Code. 15806905
Brand: Wilmad-LabGlass™ 666090825
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Wilmad-LabGlass Benchtop NMR Tubes have been tested to assure performance and permit confidence in purchasing consumables for instruments.
  • 5mm O.D. tubes available in 7” or 8” lengths in quantities of 25 or 150 pieces
  • Packaging allows for easy tube access and storage
  • Attractively priced for a high-throughput laboratory environment
  • Type 1, Class B Borosilicate glass construction with disposable caps
Specifications
For Use With (Equipment) Benchtop NMR Spectrometers
Length (Metric) 20.3 cm
Material Borosilicate Glass
Product Type NMR Tube
Color Clear
Closure Type Cap
Diameter (Metric) Outer 5 mm
For Use With (Application) NMR Experiments
Length (English) 8 in.
Thickness (Metric) Wall 0.43 mm
Quantity 25
Model WG-BTNMR-8-25