Learn More
Thermo Scientific™ Benchtop Muffle Furnaces Accessories
Product Code. 11806695 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Hearth Tray
Length (English):
6 in.
10 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Length (English) Unit Size
11806695 Hearth Tray 10 in. Each
11800830 Hearth Tray 6 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11806695 Supplier Thermo Scientific™ Supplier No. PH480X1
  • ONLINE EXCLUSIVE PRICE
    73.75€
    70.50€ / Pack of 1
    Save 3.25€
    4% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Use these accessories with Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces.
Use Thermo Scientific™ Benchtop Muffle Furnaces Accessories with Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25.4 x 19.3 x 0.95 cm
Temperature (Metric) 100°C to 1200°C
Type Hearth Tray
Dimensions (L x W x H) Exterior 10 x 7.6 x 0.38 in. (25.4 x 19.3 x 0.95 cm)
Length (English) 10 in.
Length (Metric) 25.4 cm
Width (English) 7.6 in.
Width (Metric) 19.3 cm
Height (English) 0.38 in.
Height (Metric) 0.95 cm
Voltage 0/1 V