Learn More
General Tools Benchtop Tripod for Sound and Light Meters
Product Code. 11775748 Shop All Cole-Parmer Products
Click to view available options
Height (Metric):
8.9 cm
Unit Size:
Each
Product Code. 11775748
Brand: General Tools 0161851
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use the Benchtop Tripod for Sound and Light Meters to keep your hands free or use for extended monitoring.
  • 5 year warranty
Specifications
Height (Metric) 8.9 cm
Height (English) 3.5 in.
Includes Integral Tripod Thread, Remote Sensor With Cover, Carrying Case, and One 9V Battery
Product Type Light Meter Tripod/stand
For Use With (Application) For Sound and Light Meters
Quantity 1 Ea.