Learn More
Thermo Scientific™ Benchtop Coolers
BIOPHARMA
Product Code. p-4520724
Change view
Click to view available options
Temperature Range:
-20°C
0°C
Array:
3 x 4
4 x 5
4 x 8
Holds:
12 Cryogenic Vials (0.2 to 1.5 mL)
12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
12 Tubes (0.2 to 0.5 mL)
12 Tubes (12 to 13 mm dia.)
12 Tubes (16 to 17 mm dia.)
20 Tubes (0.2 to 0.5 mL)
32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
32 Tubes (0.2 to 0.5 mL)
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Array Holds unitSize
10777232 -20°C 4 x 5 20 Tubes (0.2 to 0.5 mL) Each
10292581 -20°C 3 x 4 12 Tubes (0.2 to 0.5 mL) Each
10252631 -20°C 4 x 8 32 Tubes (0.2 to 0.5 mL) Each
10525021 0°C 3 x 4 12 Tubes (12 to 13 mm dia.) Each
10253921 0°C 3 x 4 12 Tubes (16 to 17 mm dia.) Each
10089140 0°C 3 x 4 12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL) Each
10778391 0°C 4 x 8 32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL) Each
10741792 -20°C 3 x 4 12 Cryogenic Vials (0.2 to 1.5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10777232 Supplier Thermo Scientific™ Supplier No. 355501
Only null left
Add to basket
Add to basket
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 133 mm
Height (English) 8.25 in.
Length (Metric) 226 mm
Length (English) 8.94 in.
Width (Metric) 165 mm
Width (English) 6.5 in.
Dimensions (L x W x H) 8.94 x 6.5 x 8.25 in. (226 x 165 x 133 mm)
Description 4x5 compartments, -20 deg.
Array 4 x 5
Holds Temp., Hours <-15°C, 1
Design Non-filled/clear lid
Material Polycarbonate
Type Benchtop Cooler
Color Clear
Capacity (Metric) 0.2 to 0.5 mL
Holds 20 Tubes (0.2 to 0.5 mL)
Temperature Range -20°C