Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Product Code. 16042042
Click to view available options
For Use With (Application):
ÄKTA Avant
ÄKTA Pure
ÄKTA Start
No. per Pack:
10/Pk.
25/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 25
Pack of 50
Product Code. 16042042
Brand: Cytiva 230010074
Protects the top buffer trays from buffer spillages and salt deposits
  • Cover the top buffer tray to protect from buffer spillages and salt deposits
  • Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
  • Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
  • Smooth, absorbent Whatman paper quickly absorbs spills
  • Laminated polyethylene layer prevents flow-through to the working surface
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Convenient and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Absorbent Liner
Material Paper, Laminated Polyethylene
For Use With (Application) ÄKTA Avant
No. per Pack 50/Pk.