Learn More
Wheaton™ Bench-Top Small Bottle Roller Apparatus
Product Code. 10748771 Shop All Wheaton Science Products Products
Change view
Click to view available options
Model:
C Model
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10748771 C Model Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10748771 Supplier Wheaton™ Supplier No. W348924C
  • ONLINE EXCLUSIVE PRICE
    2163.00€
    1619.14€ / Pack of 1
    Save 543.86€
    25% Off
 Restricted Item
Estimated Shipment: 30-11-2026
For cell cultivation research and pilot applications. Wheaton™ Bench-Top Small Bottle Roller Apparatus allows for use with glass or plastic roller bottles.
  • Accommodates two bottles 75 to 121mm in diameter and 290mm long
  • Deck kits available to double or triple capacity
  • Can be operated in standard incubators and chill rooms (5° to 40°C)
  • Precise and reproducible speed adjustments
Specifications
Speed Range 0.1 to 3.8 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 14 W
Voltage 100/240 V
Temperature Range (Metric) 40°C
Wattage 14 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Depth (Metric) 32.5 cm
Height (Metric) 18 cm
Width (Metric) 32 cm
Weight (Metric) 8 kg
Model C Model
For Use With (Application) For the growth and observation of various cell cultures
Packaging Quantity 1 Ea.