Learn More
Wilmad-LabGlass™ SP Scienceware™ 3mm O.D. Thin Walled Precision NMR Tube
Product Code. 15358968 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
178 mm
203.2 mm
228.6 mm
Model:
305-PS-7-5
305-PS-8-5
305-PS-9-5
307-PP-7-5
307-PP-8-5
307-PP-9-5
327-PP-7-5
327-PP-8-5
327-PP-9-5
328-PP-7-5
328-PP-8-5
328-PP-9-5
335-PP-7-5
335-PP-8-5
335-PP-9-5
Field Strength:
200MHz
300MHz
400MHz
500MHz
600MHz
Unit Size:
Pack of 5
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Product Code. 15358968
Brand: Wilmad-LabGlass™ 665000385
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufactured in a state-of-the-art ISO 9001:2008 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor.
  • Inner surface is resistant to strong acid and base at ambient temperature
  • Can be operated safely at temperatures up to 230°C, and within a temperature step of 120°C
  • Rated as hydrolytic class 1
  • Made from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
Specifications
Camber (Metric) 13 μm
Concentricity (Metric) 25μm
Length (Metric) 178 mm
Material Glass (Type 1, Class A)
Diameter (Metric) Outer 2.9935±0.0065 mm
Diameter (Metric) Inner 2.4195±0.0065 mm
Field Strength 500MHz
Length (English) 7 in.
Thickness (Metric) Wall 0.29 mm
Model 328-PP-8-5