Learn More
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags
Product Code. 11781495 Shop All SP Scienceware Products
Click to view available options
Dimensions (LxW):
5 x 7 in.
8 x 10 in.
Unit Size:
Each
Product Code. 11781495
Brand: Bel-Art™ F133530001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
  • Made of black nylon
  • Hook-and-loop sealing strip on its closing flap
Specifications
Product Type Security Bag
Material Nylon
Color Black
For Use With (Application) Used for additional protection from light for X-ray film cassettes that you are currently using
For Use With (Equipment) X-ray Film Cassette
Dimensions (LxW) 8 x 10 in.