Learn More
Bel Art™ Wire Lead Flask with Vikem™ Coating
Product Code. 12363946 Shop All SP Scienceware Products
Click to view available options
Diameter (English) Outer:
0.25 in.
0.31 in.
Diameter (Metric) Outer:
0.64 cm
0.8 cm
Weight (English):
0.25 lb.
0.38 lb.
Weight (Metric):
114 g
176 g
Unit Size:
Each
Pack of 5
Product Code. 12363946
Brand: Bel Art™ F183240000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Wire lead weights coil into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath.
  • Can be coiled around flasks, threaded through handles or looped around bottle necks
  • Coated with chemical resistant red Vikem vinyl
  • The 30 cm (12 in.) length and flexibility make these weights very versatile
Specifications
Product Type Lead Ring Flask Weight
Autoclavable Not Autoclavable
Color Red
Holds Flasks (Assorted Sizes)
Material Vinyl-coated Lead
Positions 1
Shape Cylindrical
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) Flasks
Length (Metric) 30 cm
Width (Metric) 20 mm
Height (Metric) 20 mm
Diameter (English) Outer 0.25 in.
Diameter (Metric) Outer 0.64 cm
Diameter (Metric) Inner 20 mm
Weight (English) 0.25 lb.
Weight (Metric) 114 g