Learn More
Bel-Art™ Transpette™ Eight Channel Transfer Pipettor
Product Code. 10075210 Shop All SP Scienceware Products
Click to view available options
Sterility:
Non-sterile
Sterile
Material:
Polyethylene
Polypropylene
Packaging Type:
Bulk
Individually Wrapped
Unit Size:
Pack of 100
Pack of 25
Product Code. 10075210
Brand: Bel-Art™ F378730001
For washing plates, feeding, and supernatant aspiration from microwell plates
  • 8-Channel with 600μL capacity per channel
  • Made of translucent plastic
  • Discard after hot radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options
Specifications
Material Polyethylene
Sterility Sterile
For Use With (Application) For washing plates, feeding, and supernatant aspiration from microwell plates
Quantity 100/Cs.
Packaging Type Individually Wrapped
Color Natural
Volume (Metric) 600 μL
Length (Metric) 180 mm
Type Transfer Pipet