Learn More
Bel-Art™ Super Biohazard Disposal Bags
Product Code. 17662085
Change view
Click to view available options
Dimensions (L x W):
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
31 x 38 in. (79 x 96 cm)
35 x 25 in. (35 x 64 cm)
37 x 48 in. (94 x 122 cm)
Unit Size:
Pack of 100
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17662085 19 x 14 in. (48 x 36 cm) Pack of 200
10287083 35 x 25 in. (35 x 64 cm) Pack of 200
17672085 23 x 19 in. (58 x 48 cm) Pack of 200
17692085 37 x 48 in. (94 x 122 cm) Pack of 100
17682085 31 x 38 in. (79 x 96 cm) Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17662085 Supplier Bel-Art™ Supplier No. F131651419
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Capacity (Metric) 165 L
Closure Type Zip Tie
Capacity (English) 43.6 gal.
Color Red
Dimensions (L x W) 19 x 14 in. (48 x 36 cm)
Disposable Single-use
Length (Metric) 48 cm
Sterility Non-sterile
Width (Metric) 36 cm
Autoclavable Autoclavable
Includes Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
For Use With (Application) Extra-strength, high-impact, high-temperature bags reduce autoclave time
Width (English) 14 in.
Length (English) 19 in.
Thickness 2 mil
Material Polymer
Product Type Biohazard Disposal Bag