Description
- Fit auto and nonelectric models
- Durable, clear polystyrene construction
- Perforated to faciilitate air flow
Specifications
Specifications
| Diameter (Metric) Outer | 51.8 cm |
| Product Type | Desiccator Shelf (Vertical) |
| Material | Polystyrene |
| For Use With (Application) | Convert 4.0 units from horizontal to vertical desiccator type |