Learn More
Bel-Art™ Replacement Chucks for Pipette Pump™ Pipettors
Shop All SP Scienceware Products
Click to view available options
For Use With (Equipment):
10mL pipette pump pipettor
2mL pump pipettor
Unit Size:
Each
Description
- For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
- Pipettes fit smoothly into these flexible tapered chucks
- Replacement chucks to fit various sizes/colors of Pipette Pumps in case of loss or damage
- Silicone rubber is resistant to acids and alkalis
- Disassemble parts for cleaning; chuck should not be autoclaved
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With | Pipette Pump Pipettors |
| Material | Silicone Rubber |
| Type | Chuck |
| For Use With (Equipment) | 10mL pipette pump pipettor |