Learn More
Bel Art™ PLUM™ Emergency Eyewash Station Refills
Product Code. p-4908406 Shop All SP Scienceware Products
Click to view available options
Product Type:
Saline Eyewash Solution Refill
Includes:
Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Capacity (Metric):
500 mL
Unit Size:
Pack of 3
Product Code. 12719706
Brand: Bel Art™ F248800053
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Refill kits for PLUM station
Refill kits for PLUM station
  • Bottles contain a sterile 0.9% sodium chloride solution (saline) for rinsing dust and debris, or a 4.9% sterile phosphate buffer (pH neutralizing) solution to quickly neutralize acids and alkalis
  • Bottles have clear first aid instructions, twist-open design for quick access and a built-in eye cup with dust cap
  • Use to refill eye wash stations or take to job sites as part of first aid kits
  • Bottles require no maintenance and have a three-year shelf life from date of manufacture
Compliance
Meet all U.S. federal regulations. Comply with ANSI™ standards. NDC# and expiration date printed on every bottle.
Specifications
Capacity (English) 16 oz.
Capacity (Metric) 500 mL
For Use With (Equipment) PLUM Eyewash Stations or Alone
Includes Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Product Type Saline Eyewash Solution Refill