Learn More
Bel Art™ Magnetic Bead Separation Racks
Product Code. 16650584 Shop All SP Scienceware Products
Click to view available options
Holds:
PCR Tube Plate (96-well)
Tubes (2 mL)
Tubes (5 or 15 mL)
Unit Size:
Each
Product Code. 16650584
Brand: Bel Art™ F199000002
Only null left
Add to basket
Add to basket
Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.
  • Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves of 1.5 to 50 mL models allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
  • 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes
  • Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents
Specifications
Product Type Magnetic Bead Separation Rack
Array 2 x 5
Autoclavable Not Autoclavable
Color Clear
Holds Tubes (2 mL)
Material Neodymium Magnets, Acrylic
Positions 10
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Application) General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
Width (Metric) 102 mm
Height (Metric) 38 mm