Learn More
Bel-Art™ Large Policeman
Description
- Large, tapered, flattened end is 1 in. (25mm) wide for ease in sample handling
- For aggressive scraping and spreading
- Blister packed
Specifications
Specifications
| Autoclavable | Autoclavable |
| Shape | Straight |
| Product Type | Policemen Scraper |
| For Use With (Application) | Designed for aggressive scraping and spreading without scratching |
| Length (English) | 8 in. |
| Size | Large |
| Diameter (English) Outer | 0.37 in. |
| Diameter (Metric) Outer | 0.952 cm |
| Includes | 1 x 1 in. Wide flattened and tapered end |
| Handle Material | Steel |
| Show More |