Learn More
Bel Art™ Lab-Aire™ II Glassware Drying Racks
Product Code. p-3568847
Change view
Click to view available options
Positions:
19
38
57
76
114
Product Type:
Benchtop Rack (1-tier)
Benchtop Rack (2-tier)
Benchtop Rack (3-tier)
Benchtop/Wall-mount Rack (1-tier)
Wall-mount Rack (2-tier)
Wall-mount Rack (3-tier)
Height (English):
8.4 in.
9.4 in.
15.4 in.
16.4 in.
22.4 in.
23.4 in.
Depth (English):
5 in.
7 in.
10 in.
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Product Type Height (English) Depth (English) unitSize
11872801 76 Benchtop Rack (2-tier) 15.4 in. 10 in. Each
15540102 57 Benchtop Rack (3-tier) 22.4 in. 7 in. Each
15530102 38 Benchtop Rack (2-tier) 15.4 in. 7 in. Each
11862801 38 Benchtop Rack (1-tier) 8.4 in. 10 in. Each
11803001 19 Benchtop/Wall-mount Rack (1-tier) 9.4 in. 5 in. Each
11302754 57 Wall-mount Rack (3-tier) 23.4 in. 5 in. Each
11866331 38 Wall-mount Rack (2-tier) 16.4 in. 5 in. Each
11882801 114 Benchtop Rack (3-tier) 22.4 in. 10 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11872801 Supplier Bel Art™ Supplier No. F189330022
Only null left
Add to basket
Add to basket
Glassware and plasticware drying racks for walls or benchtops
Glassware and plasticware drying racks for walls or benchtops
  • Dry and store delicate labware including beakers, flasks, bottles, test tubes and cylinders on these space-saving drying racks
  • Single-sided benchtop models fit flat against a wall
  • Double-sided benchtop models provide double the capacity (up to 114 pegs) with a slightly larger footprint
  • Three sizes and lengths of V-channel pegs can be repositioned to hold a variety of labware
  • Rugged ABS plastic body and pegs wipe clean and won't corrode
  • Wall-mount models have mounting points that fit standard pegboards
  • Pegs available separately to reconfigure rack for specific drying requirements
Specifications
Autoclavable Not Autoclavable
Color Cream, Teal
Holds Lab Glassware, Lab Plasticware
Material ABS
Positions 76
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Lab Glassware
Size 375 x 254 x 569 mm
Length (Metric) 569 mm
Width (English) 14.75 in.
Height (English) 15.4 in.
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Includes Pegs
Product Type Benchtop Rack (2-tier)
Shelves None
Height (Metric) 39 cm
Array 76 x 1
Width (Metric) 37.5 cm