Learn More
Bel-Art™ Glove Box Economy Sleeved Gloves
Product Code. 11765084 Shop All SP Scienceware Products
Click to view available options
:
Large (size 10)
Product Code. 11765084
Brand: Bel-Art™ H500290000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Compatible with Scienceware Portable Glove Box or any glove box with 6 in. glove ports
  • Durable neoprene fabrication
  • Safeguards against gas, liquid, or solid substance penetrants
  • Use with Clamping Ring, Part No. 03-421-102
  • Large, size 10
  • 1 pair/Pk.
Specifications
Color Black
Hand Specific
Latex Free No
Material Bonded Neoprene
Product Type Glove Box Gloves
Size 10
Thickness (English) 18 mil
Thickness (Metric) 0.4572 mm