Learn More
Bel Art™ Delrin™ Spring Tubing Clamps
Product Code. p-31036 Shop All SP Scienceware Products
Click to view available options
Diameter (Metric) Inside:
8 mm
12 mm
Unit Size:
Pack of 3
Product Code. 10294441
Brand: Bel Art™ H182140000
Self-closing spring operated tubing clamp is suitable for all flexible tubing
  • Positive-actionclamp opens by squeezing handles together
  • Naturally clamps shut to stop flow
  • Made from chemical- and stain-resistant Delrin™ plastic and stainless steel
Specifications
Quantity 3
Type Spring Style Tubing Clamp
Material Acetal Plastic
For Use With (Equipment) All flexible 0.46 in. (1.16 cm) O.D. tubing
Color White
Jaw Type Angled
Diameter (Metric) Inside 12 mm