Learn More
Bel-Art™ Cloning Cylinders
Product Code. 10246182
Change view
Click to view available options
Size:
Large
Medium
Small
Small, Medium, Large (Variety)
Unit Size:
Pack of 40
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10246182 Large Pack of 50
11795176 Small, Medium, Large (Variety) Pack of 40
11717085 Small Pack of 50
10552683 Medium Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10246182 Supplier Bel-Art™ Supplier No. F378470300
  • ONLINE EXCLUSIVE PRICE
    114.00€
    112.52€ / Pack of 50
    Save 1.48€
    1% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Polystyrene cloning cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Order Info
Individual sizes supplied five bags of 10 cylinders each; Variety Pack contains four bags with 10 of each size
Specifications
Material Polystyrene
Diameter (Metric) Inside 9.5 mm
Diameter (Metric) 13 mm
Height (Metric) 11 mm
Sterility Sterile
Diameter (English) Inside 0.37 in.
Product Type Cloning Cylinder
Height (English) 0.443 in.
For Use With (Application) Individual clone isolation and propagation
Size Large