Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
Product Code. 12409620 Shop All SP Scienceware Products
Click to view available options
For Use With:
4.7 to 6mm
4.7 to 9.5mm
6 to 12.7mm
9.5 to 12.7mm
Fits 9 x 6mm Tubing I.D.
Unit Size:
Pack of 12
Product Code. 12409620
Brand: Bel-Art™ 0645840
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 0.25 to 0.375 in.
Material Polypropylene
For Use With Fits 9 x 6mm Tubing I.D.
Packaging Quantity 12 Pack
Port Size 57 x 18mm
Includes Reducing connector
Color Clear
Product Type Reducing Connector