Learn More
Bel Art™ Lab-Aire™ II Glassware Dryers Accessories
Product Code. p-4508104 Shop All SP Scienceware Products
Click to view available options
Length (English):
3 in.
4 in.
5 in.
Size:
Small
Medium
Large
Unit Size:
Pack of 19
Product Code. 13293409
Brand: Bel Art™ F189310002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Replacement pegs for use with Lab-Aire II benchtop and wallmount dryer models.
  • Available as small (3 in.), medium (4 in.) and large (5 in.) pegs
  • Pegs can be moved and repositioned to hold variety of labware including beakers, flasks, bottles, test tubes and cylinders
Specifications
Includes Pegs
Product Type Replacement Peg
Autoclavable Not Autoclavable
Color Teal
Holds Lab Glassware
Material ABS
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) Lab-Aire II benchtop and wallmount dryers
Size Medium
Length (English) 4 in.