missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ Behrotest™ Removal Glassware
Click to view available options
Width (Metric):
0.27 m
Unit Size:
Each
Description
- For use with phenol distillation
Specifications
Specifications
| Depth (Metric) | 0.57 m |
| Height (Metric) | 0.28 m |
| Width (Metric) | 0.27 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction