missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ behrotest™ Distillation Attachment

Product Code. 10771413
Click to view available options
:
ForUseWithApplication:Phenol distillation
This item is not returnable. View return policy

Product Code. 10771413

Brand: behr Labor-Technik™ B00218196

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with phenol distillation. behr Labor-Technik™ behrotest™ Distillation Attachment for use with phenol distillation.

  • Attachment for phenol distillation

Specifications

For Use With (Application) Phenol Distillation
Size Width: 0.3m, Height: 0.09m, Depth: 0.32m
Length (Metric) 0.32 m
Width (Metric) 0.3 m
Height (Metric) 0.09 m
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.