Learn More
behr Labor-Technik™ behrotest™ Dilution Water Tank
Product Code. 10460071
Change view
Click to view available options
Volume (Metric):
30 L
60 L
Diameter (Metric):
350 mm
400 mm
Includes:
Distributor frits, compressed air connection
Distributor frits, compressed air connection, internal stainless steel cooling hose and connection for recirculation thermostat
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Includes unitSize
10460071 30 L 350 mm Distributor frits, compressed air connection, internal stainless steel cooling hose and connection for recirculation thermostat Each
10516402 30 L 350 mm Distributor frits, compressed air connection Each
10065170 60 L 400 mm Distributor frits, compressed air connection Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10460071 Supplier behr Labor-Technik™ Supplier No. B00217796
Made from low-density polyethylene. behr Labor-Technik™ behrotest™ Dilution Water Tank is designed to collect waste that is discharged directly from labs via special drain lines.
  • Includes aeration setup, level display and stop tap
  • Colored (light protection), with lid for easy cleaning
Specifications
Color Blue
Volume (Metric) 30 L
Includes Distributor frits, compressed air connection, internal stainless steel cooling hose and connection for recirculation thermostat
Material LDPE
Type Dilution Water Tank
Height (Metric) 500 mm
Diameter (Metric) 350 mm