Learn More
behr Labor-Technik™ behropur™ Water Deionizer Made Of Nylon
Click to view available options
Flow Rate:
300 L/h
500 L/h
500 L/hr.
800 L/h
Capacity:
1200 L
2400 L
5500 L
Unit Size:
Each
Description
- Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
- Can be connected to the raw water network directly and without a pressure reducer
- Resistant to alternating pressure
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Also available with shutoff when the limited value is reached and limit control
- Pressure resistant to 8 bar
- Also available with shutoff when the limited value is reached and limit control
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Resistant to alternating pressure
- Can be connected to the raw water network directly and without a pressure reducer
Specifications
Specifications
| Description | B22DNZ Pressure resistant mixed-bed unit made of nylon, permanent pressure resistant to 8 bar, spare cartridge |
| Capacity | 2400 L |
| Flow Rate | 500 L/h |
| Material | Nylon |
| Model | B22dNZ |
| Type | Demineralizers |