missing translation for 'onlineSavingsMsg'
Learn More

Behentrimonium Chloride (>80%), TRC

Product Code. 30335724
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335724 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335724 Supplier Toronto Research Chemicals Supplier No. TRCB100100500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Docosyltrimethylammonium Chloride
CAS 17301-53-0
Molecular Formula C25 H54 N . Cl
Quantity 500 mg
Synonym ,N,N-Trimethyldocosan-1-aminium Chloride, Arquad 22-80, Behenyltrimethylammonium Chloride, Catinal DC 80, Cation VB 800, Docosyltrimethylammonium chloride, Econol TM 22, Gemanin KDM-P, Genamin DDMP, Genamin GDMP, Incroquat Behenyl TMC, Incroquat Behenyl TMC 85, Incroquat TMC 95, Kemamine Q 2803C, N-Docosyl-N,N,N-trimethylammonium chloride, Neoscoap CN 30SF, Nissan Cation VB, Quartamin 2285E-E, Trimethylbehenylammonium chloride, Varisoft BT 85
InChI Formula InChI=1S/C25H54N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(2,3)4;/h5-25H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name docosyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 404.156
Formula Weight 403.394
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.