Learn More
Kartell™ Beaker low form with permanent blue graduations PMP 50mL
Accurately measures the volume of a liquid. Kartell™ Low Form Beaker with Blue Graduations uses horizontal marked lines to assist in measuring out various volumes of a liquid.
Brand:  Kartell™ 0172100
Product Code. 10448652
  • ONLINE EXCLUSIVE PRICE
    101.00€
    88.06€ / Pack of 20
    Save 12.94€
    12% Off
Only null left
Estimated Shipment: 16-03-2026
Description
Accurately measures the volume of a liquid. Kartell™ Low Form Beaker with Blue Graduations uses horizontal marked lines to assist in measuring out various volumes of a liquid.
  • Good chemical resistance
  • Permanent blue graduations moulded into beaker
  • Can withstand temperatures up to 170°C for short periods
  • Suitable for applications with food