Learn More
BUCHI Beaker and Drying Flasks for Rotary Evaporators
Product Code. 15520590 Shop All Buchi Products
Click to view available options
Type:
Beaker Flask
Drying Flask
Volume (Metric):
500 mL
1500 mL
4000 mL
Unit Size:
Each
Product Code. 15520590
Brand: BUCHI 11063160
Only null left
Add to basket
Add to basket
Special shaped flasks for large samples and powder drying.
  • For use with the rotary evaporators
  • Available in multiple sizes with different standard joints
Specifications
Material Borosilicate Glass (3.3)
Type Drying Flask
Volume (Metric) 1500 mL