Learn More
Pinnacle Brush™ Beaker Brushes
Product Code. 12398318
Change view
Click to view available options
Length (Metric):
26 cm
27 cm
36.5 cm
39 cm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12398318 26 cm Pack of 10
12811934 27 cm Pack of 10
12821934 39 cm Pack of 10
12851934 36.5 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12398318 Supplier Pinnacle Brush™ Supplier No. T2382
  • ONLINE EXCLUSIVE PRICE
    25.44€
    21.76€ / Pack of 10
    Save 3.68€
    14% Off
Estimated Shipment: 17-12-2026
Constructed on galvanised steel wire handle. Pinnacle Brush™ Beaker Brushes have nylon bristles for gentle cleaning.
  • Galvanized steel handle
  • Nylon bristles
Specifications
Product Type Bottle Brush
Handle Material Metal
Diameter (Metric) 3.5 cm
Length (Metric) 26 cm
Length (Metric) Brush 9.5 cm
Length (Metric) Handle 16.5 cm
Quantity 10 Pk.
Size Brush Circumference: 11 cm