Learn More
Gravis Beaded Rim Bottles
Click to view available options
Volume (Metric):
13 mL
96 mL
Unit Size:
Pack of 210
Pack of 86
Description
- Transparent glass
- Polyethylene snap cap
Specifications
Specifications
| Color | Transparent |
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| Disposable | Reusable |
| Volume (Metric) | 96 mL |
| Graduated | No |
| Includes | Snap cap |
| Closure Material | Polyethylene |
| Autoclavable | Autoclavable |
| Closure Type | Snap Cap |
| Show More |