Learn More
Lovibond™ BD606 BOD respirometric system
Product Code. 15328314
Change view
Click to view available options
Type:
BOD Respirometric System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15328314 BOD Respirometric System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15328314 Supplier Lovibond™ Supplier No. 2444465
  • ONLINE EXCLUSIVE PRICE
    4835.00€
    4703.70€ / Pack of 1
    Save 131.30€
    2% Off
Estimated Shipment: 26-10-2026
includes 2 x BD600 Includes 2 x BD600 control unit with 6 sensor heads each 12 bottles gaskets and stir bars 2 overflow flasks nitrification inhibitor and KOH
The BD 600 is a respirometric system for the determination of Biochemical Oxygen Demand (BOD). Save time, reduce the potential for errors and easily interpret data to help make process control decisions for your plant.
The BD 600 is the ideal system for process control or as a secondary test to the Dilution Method. Using a respirometric system solves many of the issues associated with the dilution method for BOD analysis.
Significantly Reduced Sample Preparation
It's easy to begin collecting data - simply add sample to the bottle, add nitrification inhibitor and place the sensor head on the bottle. There is no need for sample dilutions or seeding the sample. Because this is a respirometric test, concerns regarding air in the sample are eliminated.
Operators do not have to be present at completion of test
Measurements are automatically taken and stored at regular time intervals and can be recalled at your convenience - no more weekend trips to the lab or plant!
Easy and quick interpretation of measurement values
In addition to displaying results of a sample directly in mg/l BOD, the large, integrated, display is able to display the BOD curve which makes it is easy to see and understand measurement data points and trends. In addition, each sensor head has an integrated LED. This allows operators to visually confirm which sample the data is being displayed for and reduces potential confusion or errors over which sample data is presented
Consistent Measurement Conditions
The Automatic Start Function ensures all measurements are taken at an equal sample temperature. When the measurement conditions are consistent across readings, operators do not have to work about temperature or pressure fluctuations skewing results.
Sample Identification and Tracking
The BD 600 interface, makes it easy to assign an identification code or sample name when setting up a new sample. This helps track a sample from the time it is collected and ensures traceability during sample measurement.
Use the remote control
If using an incubator with a glass door, operators can use the supplied remote control to display measurement data for a sample - without even opening the door! This further ensures that samples stay at a consistent temperature which is essential to accurate results.
Specifications
Type BOD Respirometric System
For Use With (Application) Determination of Biochemical Oxygen Demand or as a Secondary Test to The Dilution Method