Learn More
BD Green Nucleic Acid Stain
Product Code. 15884529 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884529 0.1 mL Each
15814539 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15884529 Supplier BD Supplier No. 565799
 Restricted Item
Estimated Shipment: 17-11-2026
Useful for discrimination of viable from nonviable cells, DNA content analysis in fixed cells, or nuclear counterstaining for fixed cells in immunofluorescence or flow cytometry applications. BD Via-Probe™ Green Nucleic Acid Stain brightly stains nonviable or fixed cells with permeable membranes.
  • Green Nucleic Acid Stain has an excitation maximum of 503nm, and is best excited by the blue laser
  • It has an emission maximum of 526nm
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Quantity 0.1 mL
For Use With (Application)
  • Immunofluorescence
  • Immunocytochemistry
  • Flow cytometry
  • Intracellular staining (flow cytometry)
Color Green
Product Type Green Nucleic Acid Stain