Learn More
BD SafetyGlide™ Insulin, TB and Allergy Syringe with Needle
Click to view available options
Needle Gauge:
31 G
Unit Size:
Pack of 400
Description
Tiny Needle Technology completes the BD SafetyGlide line to meet your Sub-Q and intradermal injection need. Includes safety shield.
Specifications
Specifications
| Needle Gauge | 31 G |
| Length (English) Needle | 0.312 in. |
| Product Type | Insulin Syringe |
| Volume (Metric) | 0.3 mL |
| Sterility | Sterile |