Learn More
BD Vacutainer BD Vacutainer™ EST Tube
Description
- Evacuated secondary tube (EST)
- No additives
- Suitable as a secondary tube for anti-coagulated blood samples, for example for taking plasma samples from blood bags
- Can also be used as a discard tube
Specifications
Specifications
| Additive | No Additive |
| Material | PET |
| Capacity (Metric) | 11 mL |
| Tube Size | 16 x 100 mm |
| For Use With (Application) | Secondary tube for anti-coagulated blood samples. |
| Label Type | Paper |