missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Rat TNF ELISA Kit
Click to view available options
Quantity:
2 Plates
Unit Size:
Each
Description
- Solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)
- Utilizes a monoclonal antibody specific for rat TNF coated on a 96-well plate
Specifications
Specifications
| Antigen | TNF |
| Cross Reactivity | Rat |
| Includes | Stop solution, standard/Sample diluent, TMB one-step Substrate reagent, PerCP-Cy™5.5 Mouse Anti-Human CD13, Enzyme diluent, rat TNF Antibody coated plate, rat TNF standard, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), det |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Quantitative determination of TNF, ELISA |
| Product Type | Rat TNF ELISA kit |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction