missing translation for 'onlineSavingsMsg'
Learn More

BD Rat Immunoglobulin Isotyping ELISA Kit

Product Code. 15828838
Change view
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15828838 80 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15828838 Supplier BD Supplier No. 557081
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

8 rat immunoglobulin isotype-specific mouse monoclonal antibodies, Horseradish peroxidase (HRP)-conjugated mouse anti-rat Ig antibody, Substrate and Stop Solutions, Coating Buffer and Sample Diluent, Positive control antigen mixture

Monoclonal antibody-based kit for identification of immunoglobulin isotypes

  • Employs a direct horseradish peroxidase-labeled system
  • Assay format eliminates need for coating the plate with antigen

Applications:

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Cross Reactivity Rat
Includes 0.25mL Monoclonal Mouse Anti-rat IgG1, 0.25mL Monoclonal Mouse Anti-rat IgG2a, 0.25mL Monoclonal Mouse Anti-rat IgG2b, 0.25mL Monoclonal Mouse Anti-rat IgG2c, 0.25mL Monoclonal Mouse Anti-rat IgM, 0.25mL Monoclonal Mouse Anti-rat IgA, 0.25mL Monoclonal Mouse Anti-rat Igkappa light chain, 0.25mL positive reference Antigen mixture, 0.25mL Monoclonal Mouse Anti-rat Ig lambda light chain, 1mL HRP Mouse Anti-rat Ig MAB, 40mL PBS Buffer (10X), 30mL BSA solution (10%), 40mL Substrate reagent A, 40mL Substrate reagent B, 40mL stop solution
Quantity 80 Tests
Content And Storage 4°C
For Use With (Application) Identification of Immunoglobulin isotypes, ELISA
Product Type Rat Immunoglobulin isotyping ELISA kit

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.